Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 17 - 18 | 1833 | 68 | |
| XM_007650653.1 | XP_007648843.1 | 18 - 19 | 1855 | 68 | ||
| XM_007650654.1 | XP_007648844.1 | 17 - 18 | 1685 | 68 | ||
| XM_007650655.1 | XP_007648845.1 | 16 - 17 | 1790 | 68 | ||
| XM_007650656.1 | XP_007648846.1 | 16 - 17 | 1754 | 68 | ||
| XM_007650657.1 | XP_007648847.1 | 17 - 18 | 1867 | 68 | ||
| XM_007650658.1 | XP_007648848.1 | 16 - 17 | 1779 | 68 | ||
| XM_007650659.1 | XP_007648849.1 | 15 - 16 | 1711 | 68 | ||
| XM_007650660.1 | XP_007648850.1 | 14 - 15 | 1585 | 68 | ||
| XM_007650661.1 | XP_007648851.1 | 10 - 11 | 1276 | 68 | ||
| GenBank mRNA | JI885650.1 | - | 7 - 8 | 637 | 68 | |
| JP050903.1 | - | 7 - 8 | 637 | 68 | ||
More Information
Set Membership: |
|

LOC100755304
Genomic Map