Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 19 - 20 | 2032 | 64 | |
| XM_007650653.1 | XP_007648843.1 | 20 - 21 | 2054 | 64 | ||
| XM_007650654.1 | XP_007648844.1 | 19 - 20 | 1884 | 64 | ||
| XM_007650655.1 | XP_007648845.1 | 18 - 19 | 1989 | 64 | ||
| XM_007650656.1 | XP_007648846.1 | 18 - 19 | 1953 | 64 | ||
| XM_007650657.1 | XP_007648847.1 | 19 - 20 | 2066 | 64 | ||
| XM_007650658.1 | XP_007648848.1 | 18 - 19 | 1978 | 64 | ||
| XM_007650659.1 | XP_007648849.1 | 17 - 18 | 1910 | 64 | ||
| XM_007650660.1 | XP_007648850.1 | 16 - 17 | 1784 | 64 | ||
| XM_007650661.1 | XP_007648851.1 | 12 - 13 | 1475 | 64 | ||
| GenBank mRNA | JI885650.1 | - | 9 - 10 | 836 | 64 | |
| JP050903.1 | - | 9 - 10 | 836 | 64 | ||
More Information
Set Membership: |
|

LOC100755304
Genomic Map