Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 20 - 21 | 2206 | 75 | |
| XM_007650653.1 | XP_007648843.1 | 21 - 22 | 2228 | 75 | ||
| XM_007650654.1 | XP_007648844.1 | 20 - 21 | 2058 | 75 | ||
| XM_007650655.1 | XP_007648845.1 | 19 - 20 | 2163 | 75 | ||
| XM_007650656.1 | XP_007648846.1 | 19 - 20 | 2127 | 75 | ||
| XM_007650657.1 | XP_007648847.1 | 20 - 21 | 2240 | 75 | ||
| XM_007650658.1 | XP_007648848.1 | 19 - 20 | 2152 | 75 | ||
| XM_007650659.1 | XP_007648849.1 | 18 - 19 | 2084 | 75 | ||
| XM_007650660.1 | XP_007648850.1 | 17 - 18 | 1958 | 75 | ||
| XM_007650661.1 | XP_007648851.1 | 13 - 14 | 1649 | 75 | ||
| GenBank mRNA | JI885650.1 | - | 10 - 11 | 1010 | 75 | |
| JP050903.1 | - | 10 - 11 | 1010 | 75 | ||

LOC100755304
Genomic Map