Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 21 - 22 | 2301 | 103 | |
| XM_007650653.1 | XP_007648843.1 | 22 - 23 | 2323 | 103 | ||
| XM_007650654.1 | XP_007648844.1 | 21 - 22 | 2153 | 103 | ||
| XM_007650655.1 | XP_007648845.1 | 20 - 21 | 2258 | 103 | ||
| XM_007650656.1 | XP_007648846.1 | 20 - 21 | 2222 | 103 | ||
| XM_007650657.1 | XP_007648847.1 | 21 - 22 | 2335 | 103 | ||
| XM_007650658.1 | XP_007648848.1 | 20 - 21 | 2247 | 103 | ||
| XM_007650659.1 | XP_007648849.1 | 19 - 20 | 2179 | 103 | ||
| XM_007650660.1 | XP_007648850.1 | 18 - 19 | 2053 | 103 | ||
| XM_007650661.1 | XP_007648851.1 | 14 - 15 | 1744 | 103 | ||
| GenBank mRNA | JI885650.1 | - | 11 - 12 | 1105 | 103 | |
| JP050903.1 | - | 11 - 12 | 1105 | 103 | ||
More Information
Set Membership: |
|

LOC100755304
Genomic Map