Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 8 - 9 | 694 | 96 | |
| XM_007650653.1 | XP_007648843.1 | 9 - 10 | 716 | 96 | ||
| XM_007650654.1 | XP_007648844.1 | 8 - 9 | 546 | 96 | ||
| XM_007650655.1 | XP_007648845.1 | 7 - 8 | 651 | 96 | ||
| XM_007650656.1 | XP_007648846.1 | 7 - 8 | 615 | 96 | ||
| XM_007650657.1 | XP_007648847.1 | 8 - 9 | 728 | 96 | ||
| XM_007650658.1 | XP_007648848.1 | 7 - 8 | 640 | 96 | ||
| XM_007650659.1 | XP_007648849.1 | 6 - 7 | 572 | 96 | ||

LOC100755304
Genomic Map