Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 10 - 11 | 952 | 81 | |
| XM_007650653.1 | XP_007648843.1 | 11 - 12 | 974 | 81 | ||
| XM_007650654.1 | XP_007648844.1 | 10 - 11 | 804 | 81 | ||
| XM_007650655.1 | XP_007648845.1 | 9 - 10 | 909 | 81 | ||
| XM_007650656.1 | XP_007648846.1 | 9 - 10 | 873 | 81 | ||
| XM_007650657.1 | XP_007648847.1 | 10 - 11 | 986 | 81 | ||
| XM_007650658.1 | XP_007648848.1 | 9 - 10 | 898 | 81 | ||
| XM_007650659.1 | XP_007648849.1 | 8 - 9 | 830 | 81 | ||
| XM_007650660.1 | XP_007648850.1 | 7 - 8 | 704 | 81 | ||
| XM_007650661.1 | XP_007648851.1 | 3 - 4 | 395 | 81 | ||
| GenBank mRNA | JI870221.1 | - | 7 - 8 | 646 | 81 | |
| JP064617.1 | - | 7 - 8 | 646 | 81 | ||

LOC100755304
Genomic Map