Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 11 - 12 | 1208 | 103 | |
| XM_007650653.1 | XP_007648843.1 | 12 - 13 | 1230 | 103 | ||
| XM_007650654.1 | XP_007648844.1 | 11 - 12 | 1060 | 103 | ||
| XM_007650655.1 | XP_007648845.1 | 10 - 11 | 1165 | 103 | ||
| XM_007650656.1 | XP_007648846.1 | 10 - 11 | 1129 | 103 | ||
| XM_007650657.1 | XP_007648847.1 | 11 - 12 | 1242 | 103 | ||
| XM_007650658.1 | XP_007648848.1 | 10 - 11 | 1154 | 103 | ||
| XM_007650659.1 | XP_007648849.1 | 9 - 10 | 1086 | 103 | ||
| XM_007650660.1 | XP_007648850.1 | 8 - 9 | 960 | 103 | ||
| XM_007650661.1 | XP_007648851.1 | 4 - 5 | 651 | 103 | ||
More Information
Set Membership: |
|

LOC100755304
Genomic Map