Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 12 - 13 | 1345 | 90 | |
| XM_007650653.1 | XP_007648843.1 | 13 - 14 | 1367 | 90 | ||
| XM_007650654.1 | XP_007648844.1 | 12 - 13 | 1197 | 90 | ||
| XM_007650655.1 | XP_007648845.1 | 11 - 12 | 1302 | 90 | ||
| XM_007650656.1 | XP_007648846.1 | 11 - 12 | 1266 | 90 | ||
| XM_007650657.1 | XP_007648847.1 | 12 - 13 | 1379 | 90 | ||
| XM_007650658.1 | XP_007648848.1 | 11 - 12 | 1291 | 90 | ||
| XM_007650659.1 | XP_007648849.1 | 10 - 11 | 1223 | 90 | ||
| XM_007650660.1 | XP_007648850.1 | 9 - 10 | 1097 | 90 | ||
| XM_007650661.1 | XP_007648851.1 | 5 - 6 | 788 | 90 | ||
| GenBank mRNA | JI885650.1 | - | 2 - 3 | 149 | 90 | |
| JP050903.1 | - | 2 - 3 | 149 | 90 | ||

LOC100755304
Genomic Map