Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 13 - 14 | 1483 | 90 | |
| XM_007650653.1 | XP_007648843.1 | 14 - 15 | 1505 | 90 | ||
| XM_007650654.1 | XP_007648844.1 | 13 - 14 | 1335 | 90 | ||
| XM_007650655.1 | XP_007648845.1 | 12 - 13 | 1440 | 90 | ||
| XM_007650656.1 | XP_007648846.1 | 12 - 13 | 1404 | 90 | ||
| XM_007650657.1 | XP_007648847.1 | 13 - 14 | 1517 | 90 | ||
| XM_007650658.1 | XP_007648848.1 | 12 - 13 | 1429 | 90 | ||
| XM_007650659.1 | XP_007648849.1 | 11 - 12 | 1361 | 90 | ||
| XM_007650660.1 | XP_007648850.1 | 10 - 11 | 1235 | 90 | ||
| XM_007650661.1 | XP_007648851.1 | 6 - 7 | 926 | 90 | ||
| GenBank mRNA | JI885650.1 | - | 3 - 4 | 287 | 90 | |
| JP050903.1 | - | 3 - 4 | 287 | 90 | ||

LOC100755304
Genomic Map