Assay Details
| Gene Symbol: LOC100755304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650652.1 | XP_007648842.1 | 14 - 15 | 1552 | 102 | |
| XM_007650653.1 | XP_007648843.1 | 15 - 16 | 1574 | 102 | ||
| XM_007650654.1 | XP_007648844.1 | 14 - 15 | 1404 | 102 | ||
| XM_007650655.1 | XP_007648845.1 | 13 - 14 | 1509 | 102 | ||
| XM_007650656.1 | XP_007648846.1 | 13 - 14 | 1473 | 102 | ||
| XM_007650657.1 | XP_007648847.1 | 14 - 15 | 1586 | 102 | ||
| XM_007650658.1 | XP_007648848.1 | 13 - 14 | 1498 | 102 | ||
| XM_007650659.1 | XP_007648849.1 | 12 - 13 | 1430 | 102 | ||
| XM_007650660.1 | XP_007648850.1 | 11 - 12 | 1304 | 102 | ||
| XM_007650661.1 | XP_007648851.1 | 7 - 8 | 995 | 102 | ||
| GenBank mRNA | JI885650.1 | - | 4 - 5 | 356 | 102 | |
| JP050903.1 | - | 4 - 5 | 356 | 102 | ||
More Information
Set Membership: |
|

LOC100755304
Genomic Map