Assay Details
| Gene Symbol: Sftpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496162.2 | XP_003496210.1 | 2 - 3 | 200 | 63 | |
| XM_003496163.2 | XP_003496211.1 | 2 - 3 | 200 | 63 | ||
More Information
Set Membership: |
|

Sftpc
Genomic Map