Assay Details
| Gene Symbol: Gpd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496253.2 | XP_003496301.1 | 4 - 5 | 632 | 57 | |
| GenBank mRNA | JP051622.1 | - | 1 - 2 | 57 | ||
More Information
Set Membership: |
|

Gpd1l
Genomic Map