Assay Details
| Gene Symbol: Igfbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498997.2 | XP_003499045.1 | 1 - 2 | 336 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp5| Gene Symbol: Igfbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498997.2 | XP_003499045.1 | 1 - 2 | 336 | 67 | |
Set Membership: |
|