Assay Details
| Gene Symbol: LOC100772154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502156.2 | XP_003502204.1 | 6 - 7 | 985 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100772154| Gene Symbol: LOC100772154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502156.2 | XP_003502204.1 | 6 - 7 | 985 | 59 | |
Set Membership: |
|