Assay Details
| Gene Symbol: Elf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513801.2 | XP_003513849.1 | 5 - 6 | 874 | 65 | |
| XM_007654010.1 | XP_007652200.1 | 4 - 5 | 766 | 65 | ||
More Information
Set Membership: |
|

Elf1
Genomic Map