Assay Details
Gene Symbol: LOC100764781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649318.1 | XP_007647508.1 | 2 - 3 | 597 | 57 | |
XM_007649319.1 | XP_007647509.1 | 2 - 3 | 1033 | 57 | ||
XM_007649320.1 | XP_007647510.1 | 2 - 3 | 552 | 57 | ||
XM_007649321.1 | XP_007647511.1 | 2 - 3 | 988 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
