Assay Details
| Gene Symbol: Bre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496573.1 | XP_003496621.1 | 11 - 12 | 1137 | 70 | |
| XM_007651350.1 | XP_007649540.1 | 11 - 12 | 1360 | 70 | ||
| XM_007651352.1 | XP_007649542.1 | 9 - 10 | 920 | 70 | ||
| GenBank mRNA | JI892818.1 | - | 10 - 11 | 1107 | 70 | |
More Information
Set Membership: |
|

Bre
Genomic Map