Assay Details
| Gene Symbol: Snupn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497920.2 | XP_003497968.1 | 2 - 3 | 980 | 70 | |
| GenBank mRNA | JI868488.1 | - | 2 - 3 | 224 | 70 | |
| JP048578.1 | - | 2 - 3 | 224 | 70 | ||
More Information
Set Membership: |
|

Snupn
Genomic Map