Assay Details
| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498798.1 | XP_003498846.1 | 7 - 8 | 928 | 73 | |
| XM_003498799.1 | XP_003498847.1 | 8 - 9 | 937 | 73 | ||
| XM_003498800.1 | XP_003498848.1 | 6 - 7 | 808 | 73 | ||
| XM_007633259.1 | XP_007631449.1 | 7 - 8 | 946 | 73 | ||
| XM_007633271.1 | XP_007631461.1 | 4 - 5 | 643 | 73 | ||
| XM_007633279.1 | XP_007631469.1 | 5 - 6 | 649 | 73 | ||
More Information
Set Membership: |
|

Spock3
Genomic Map