Assay Details
| Gene Symbol: Kpna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501859.2 | XP_003501907.1 | 9 - 10 | 1649 | 70 | |
| GenBank mRNA | JI868315.1 | - | 9 - 10 | 1649 | 70 | |
| JP044182.1 | - | 9 - 10 | 1649 | 70 | ||
More Information
Set Membership: |
|

Kpna2
Genomic Map