Assay Details
| Gene Symbol: LOC100765926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504253.2 | XP_003504301.1 | 3 - 4 | 465 | 88 | |
| XM_007644808.1 | XP_007642998.1 | 3 - 4 | 440 | 88 | ||
| XM_007644809.1 | XP_007642999.1 | 3 - 4 | 414 | 88 | ||
More Information
Set Membership: |
|

LOC100765926
Genomic Map