Assay Details
| Gene Symbol: Pla2g5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506897.2 | XP_003506945.1 | 3 - 4 | 314 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g5| Gene Symbol: Pla2g5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506897.2 | XP_003506945.1 | 3 - 4 | 314 | 57 | |
Set Membership: |
|