Assay Details
| Gene Symbol: Uprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506986.2 | XP_003507034.1 | 1 - 2 | 534 | 68 | |
| GenBank mRNA | JI884897.1 | - | 1 - 2 | 434 | 68 | |
| JP064022.1 | - | 1 - 2 | 434 | 68 | ||
More Information
Set Membership: |
|

Uprt
Genomic Map