Assay Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509484.2 | XP_003509532.1 | 8 - 9 | 1052 | 57 | |
| GenBank mRNA | JI877257.1 | - | 8 - 9 | 1035 | 57 | |
| JP065875.1 | - | 8 - 9 | 1035 | 57 | ||
More Information
Set Membership: |
|

Cct6a
Genomic Map