Assay Details
Gene Symbol: Znf516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510385.2 | XP_003510433.1 | 3 - 4 | 3433 | 84 | |
XM_007651008.1 | XP_007649198.1 | 3 - 4 | 3539 | 84 | ||
XM_007651009.1 | XP_007649199.1 | 3 - 4 | 3599 | 84 | ||
XM_007651010.1 | XP_007649200.1 | 3 - 4 | 3533 | 84 | ||
XM_007651011.1 | XP_007649201.1 | 3 - 4 | 3428 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
