Assay Details
| Gene Symbol: Crcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512981.2 | XP_003513029.1 | 5 - 6 | 475 | 55 | |
| GenBank mRNA | JI880598.1 | - | 5 - 6 | 315 | 55 | |
More Information
Set Membership: |
|

Crcp
Genomic Map