Assay Details
| Gene Symbol: Tomm20l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515625.2 | XP_003515673.1 | 4 - 5 | 403 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tomm20l| Gene Symbol: Tomm20l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515625.2 | XP_003515673.1 | 4 - 5 | 403 | 82 | |
Set Membership: |
|