Assay Details
Gene Symbol: LOC100773492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515652.1 | XP_003515700.1 | 4 - 5 | 635 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100773492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515652.1 | XP_003515700.1 | 4 - 5 | 635 | 88 |