Assay Details
| Gene Symbol: Cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645701.1 | XP_007643891.1 | 6 - 7 | 763 | 54 | |
| GenBank mRNA | JP039348.1 | - | 2 - 3 | 330 | 54 | |
More Information
Set Membership: |
|

Cdca7
Genomic Map