Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648683.1 | XP_007646873.1 | 7 - 8 | 1291 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem180| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648683.1 | XP_007646873.1 | 7 - 8 | 1291 | 60 | |
Set Membership: |
|