Assay Details
| Gene Symbol: Vapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496754.2 | XP_003496802.1 | 2 - 3 | 227 | 134 | |
| GenBank mRNA | JP060048.1 | - | 2 - 3 | 223 | 134 | |
More Information
Set Membership: |
|

Vapb
Genomic Map