Assay Details
| Gene Symbol: Otud3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506903.2 | XP_003506951.1 | 3 - 4 | 535 | 70 | |
| GenBank mRNA | JI887498.1 | - | 3 - 4 | 527 | 70 | |
More Information
Set Membership: |
|

Otud3
Genomic Map