Assay Details
| Gene Symbol: Ddah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508477.2 | XP_003508525.1 | 1 - 2 | 302 | 80 | |
| XM_003508478.2 | XP_003508526.1 | 1 - 2 | 302 | 80 | ||
More Information
Set Membership: |
|

Ddah2
Genomic Map