Assay Details
| Gene Symbol: Bphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513194.2 | XP_003513242.1 | 3 - 4 | 435 | 64 | |
| GenBank mRNA | JI868358.1 | - | 3 - 4 | 396 | 64 | |
| JP053915.1 | - | 3 - 4 | 394 | 64 | ||
More Information
Set Membership: |
|

Bphl
Genomic Map