Assay Details
| Gene Symbol: Ckap2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007618321.1 | XP_007616511.1 | 9 - 10 | 2038 | 69 | |
| GenBank mRNA | JI882454.1 | - | 2 - 3 | 92 | 69 | |
| JP060784.1 | - | 2 - 3 | 92 | 69 | ||
More Information
Set Membership: |
|

Ckap2l
Genomic Map