Assay Details
| Gene Symbol: LOC100752705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007639527.1 | XP_007637717.1 | 7 - 8 | 947 | 85 | |
| XM_007639536.1 | XP_007637726.1 | 7 - 8 | 947 | 85 | ||
More Information
Set Membership: |
|

LOC100752705
Genomic Map