Assay Details
| Gene Symbol: Twistnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654788.1 | XP_007652978.1 | 3 - 4 | 450 | 90 | |
| GenBank mRNA | JI883742.1 | - | 2 - 3 | 383 | 90 | |
More Information
Set Membership: |
|

Twistnb
Genomic Map