Assay Details
| Gene Symbol: Chmp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653239.1 | XP_007651429.1 | 4 - 5 | 713 | 65 | |
| GenBank mRNA | JI879768.1 | - | 2 - 3 | 324 | 65 | |
More Information
Set Membership: |
|

Chmp2b
Genomic Map