Assay Details
| Gene Symbol: Glce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498933.2 | XP_003498981.1 | 3 - 4 | 719 | 68 | |
| XM_007635562.1 | XP_007633752.1 | 4 - 5 | 789 | 68 | ||
| XM_007635566.1 | XP_007633756.1 | 5 - 6 | 1013 | 68 | ||
| XM_007635575.1 | XP_007633765.1 | 4 - 5 | 929 | 68 | ||
More Information
Set Membership: |
|

Glce
Genomic Map