Assay Details
| Gene Symbol: LOC100758930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499079.2 | XP_003499127.1 | 4 - 5 | 1195 | 83 | |
| XM_007638093.1 | XP_007636283.1 | 4 - 5 | 1342 | 83 | ||
| XM_007638097.1 | XP_007636287.1 | 4 - 5 | 1204 | 83 | ||

LOC100758930
Genomic Map