Assay Details
| Gene Symbol: LOC100758930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499079.2 | XP_003499127.1 | 5 - 6 | 1670 | 82 | |
| XM_007638093.1 | XP_007636283.1 | 5 - 6 | 1817 | 82 | ||
| XM_007638097.1 | XP_007636287.1 | 5 - 6 | 1679 | 82 | ||
| GenBank mRNA | JI862146.1 | - | 1 - 2 | 216 | 82 | |
| JP060170.1 | - | 1 - 2 | 216 | 82 | ||

LOC100758930
Genomic Map