Assay Details
| Gene Symbol: LOC100758930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499079.2 | XP_003499127.1 | 8 - 9 | 2013 | 61 | |
| XM_007638093.1 | XP_007636283.1 | 8 - 9 | 2160 | 61 | ||
| XM_007638097.1 | XP_007636287.1 | 8 - 9 | 2022 | 61 | ||
| GenBank mRNA | JI862146.1 | - | 4 - 5 | 559 | 61 | |
| JP060170.1 | - | 4 - 5 | 559 | 61 | ||
More Information
Set Membership: |
|

LOC100758930
Genomic Map