Assay Details
| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500351.2 | XP_003500399.1 | 2 - 3 | 240 | 61 | |
| XM_007641078.1 | XP_007639268.1 | 2 - 3 | 242 | 61 | ||
More Information
Set Membership: |
|

Pcbp4
Genomic Map