Assay Details
| Gene Symbol: Nt5c1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503047.2 | XP_003503095.1 | 5 - 6 | 998 | 62 | |
| XM_007643667.1 | XP_007641857.1 | 6 - 7 | 1214 | 62 | ||
| XM_007643668.1 | XP_007641858.1 | 6 - 7 | 1172 | 62 | ||
| XM_007643669.1 | XP_007641859.1 | 6 - 7 | 1163 | 62 | ||
| XM_007643670.1 | XP_007641860.1 | 6 - 7 | 1112 | 62 | ||
| XM_007643671.1 | XP_007641861.1 | 5 - 6 | 989 | 62 | ||
| XM_007643672.1 | XP_007641862.1 | 5 - 6 | 941 | 62 | ||
More Information
Set Membership: |
|

Nt5c1b
Genomic Map