Assay Details
| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506190.2 | XP_003506238.1 | 5 - 6 | 666 | 60 | |
| GenBank mRNA | JI896095.1 | - | 5 - 6 | 604 | 60 | |
| JP040523.1 | - | 5 - 6 | 602 | 60 | ||
More Information
Set Membership: |
|

Pa2g4
Genomic Map