Assay Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507062.2 | XP_003507110.1 | 6 - 7 | 751 | 65 | |
| XM_007647620.1 | XP_007645810.1 | 6 - 7 | 750 | 65 | ||
| GenBank mRNA | JI871865.1 | - | 2 - 3 | 207 | 65 | |
| JP061134.1 | - | 2 - 3 | 207 | 65 | ||
More Information
Set Membership: |
|

Zfand2a
Genomic Map