Assay Details
| Gene Symbol: LOC100759356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509858.2 | XP_003509906.1 | 10 - 11 | 1162 | 115 | |
| GenBank mRNA | JI872977.1 | - | 5 - 6 | 424 | 114 | |
More Information
Set Membership: |
|

LOC100759356
Genomic Map