Assay Details
| Gene Symbol: LOC100759356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509858.2 | XP_003509906.1 | 4 - 5 | 560 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100759356| Gene Symbol: LOC100759356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509858.2 | XP_003509906.1 | 4 - 5 | 560 | 110 | |
Set Membership: |
|