Assay Details
| Gene Symbol: LOC100759356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509858.2 | XP_003509906.1 | 8 - 9 | 991 | 118 | |
| GenBank mRNA | JI872977.1 | - | 3 - 4 | 253 | 118 | |
More Information
Set Membership: |
|

LOC100759356
Genomic Map