Assay Details
| Gene Symbol: Ndufc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510390.2 | XP_003510438.1 | 1 - 2 | 322 | 127 | |
| GenBank mRNA | JI894690.1 | - | 1 - 2 | 291 | 127 | |
| JP043569.1 | - | 1 - 2 | 291 | 127 | ||
More Information
Set Membership: |
|

Ndufc2
Genomic Map